Learn More
Accuform Signs Security Signs Surveillance Systems
Communicate through informative signs where video surveillance is in use.
Supplier:  Accuform Signs MSEC563VS
 View more versions of this product
Catalog No. 19-805-234
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Non-glare
1 Ea.
Short term outdoor applications and in moderate temperatures
176°F
4 mil
-21°C to 80°C
45.7 x 30.5 cm
Security Surveillance Sign
Vinyl
Surface Mount
18 in.
-20°F
12 in.
-20°F to 176°F
Security Surveillance Systems In Operation
Product Suggestions
Product Suggestions