missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Safety Projection Sign: Emergency Shower
When you need identification, location, and direction, signs can be seen from various angles for extra visibility.
$31.88 - $34.74
Specifications
| Quantity | 1 Ea. |
|---|---|
| For Use With (Application) | Visual indicators to guide people to and mark the location of emergency showers or eyewash |
| Thickness | 10 in. |
| Product Type | Projection Sign |
| Color | White |
Description
- Hang signs along hallways, stairways, corridors and doorways
- Signs are .10 in. thick high-impact plastic
- Images on the two panels are protected by an over-laminate
- Flanged edges have pre-drilled mounting holes
- Wall mount with screws or self-adhesive foam tape (both included)
Available in two styles:
- 3D signs project 45 degrees from the wall, providing optimum viewing from the front and sides
- 90D signs project 90 degrees from the wall and can be viewed from either side
Specifications
| 1 Ea. | |
| 10 in. | |
| White |
| Visual indicators to guide people to and mark the location of emergency showers or eyewash | |
| Projection Sign |