Learn More
Accuform Signs Information Centers
Post evacuation and assembly assignments, rescue and medical duties, security measures, and more in one convenient location
Supplier:  Accuform Signs EMC224
 View more versions of this product
Catalog No. 19169550
Only null left
Description
Description
  • .063 in. Aluminum backboard
  • H x W: 20 x 15 in. (50.8 x 38.1cm)
  • Printed with bold red stripes for high visibility
  • Attached coated-wire basket with 1.5 in. dia. (3.81cm), three-ring binder
  • Four corner mounting holes for easy installation
Specifications
Chemical Identifiers
Aluminum Backboard
Safety and Training Materials
Post evacuation and assembly assignments, rescue and medical duties, security measures and more in one convenient location
Specifications
Aluminum Backboard
Safety and Training Materials
Post evacuation and assembly assignments, rescue and medical duties, security measures and more in one convenient location
Product Suggestions
Product Suggestions