missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs HazMat Warning Label Poster
Description
- Plastic laminated poster provides easy-to-understand information
- Divided into segments based on the chemical and hazard labeling system
- Defines HMCIS color bar and NFPA diamond formats along with the numerical degree of severity hazard (expressed as a numerical value)
- Also available in Spanish
Order Info
Also available in Spanish. Contact your Customer Service Representative for ordering information.
Specifications
Specifications
| Material | Plastic |
| Product Type | Hazmat Warning Label Poster |
| For Use With (Application) | Quick look up of the hazard index and personal protection |
| Includes | Safety sign, tag and label, as well as an extensive line of Lockout/Tagout products, legend identifying which personal protective equipment is necessary for a specific code |
| Dimensions (L x W x H) | 18 x 24 x 18 in. |
| Length (English) | 18 in. |
| Length (Metric) | 45.72 cm |
| Width (English) | 24 in. |
| Width (Metric) | 60.96 cm |
| Height (English) | 18 in. |
| Show More |