missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs First Aid Safety Labels

Catalog No. 17998134
Change view
Click to view available options
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17998134 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17998134 Supplier Accuform Signs Supplier No. LFSD509VSP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This First Aid Kit Inside label includes pictograms to identify location of the kit

Specifications

Material Vinyl
Quantity 5/Pk.
For Use With (Application) Labels are an easy way to provide safe and effective communication
Height (English) 3.5 in.
Thickness 4 mil
Width (English) 5 in.
Displayed Text First Aid Kit Inside
Product Type First Aid Safety Label
Color Green on White