missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Accessories for YT31042HW Hazardous Waste Filtration System, 142 mm
Accessories for filter holders for sample preparation.
Supplier: MilliporeSigma™ YT310RAHW
Includes: Accessory clamps for 1- and 2.5-liter bottle extractions
Description
Accessories for filter holders for sample preparation.
YT3014257 Underdrain Support, PTFE-coatedYT310RAHW Rotary AgitatorOrder Info
Dimensions: 21L x 26 1/4W x 24 3/8 in.H (53 x 66.6 x 61.9cm)
Specifications
| Not Autoclavable | |
| Non-sterile | |
| Accessory clamps for 1 L and 2.5 L bottle extractions | |
| Rotary Agitator |