Learn More
MilliporeSigma™ Accessories for YT31042HW Hazardous Waste Filtration System, 142 mm
Catalog No. YT3014257
Change view
Click to view available options
Type:
Rotary Agitator
Underdrain Support
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
YT3014257 Underdrain Support
YT310RAHW Rotary Agitator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. YT3014257 Supplier MilliporeSigma™ Supplier No. YT3014257
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessories for filter holders for sample preparation.
YT3014257 Underdrain Support, PTFE-coatedYT310RAHW Rotary Agitator
Specifications
Autoclavable Not Autoclavable
Material PTFE-coated Stainless Steel
Sterility Non-sterile
Type Underdrain Support
Diameter (Metric) 142 mm
For Use With (Equipment) Hazardous Waste Filtration System