Learn More
Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.
$115.00 - $6,486.61
Specifications
Product Line Varioskan
View More Specs
Products 6
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
50950611
Encompass
View Documents Promotion Details
Thermo Scientific™
N02693
96-well Adapter for Plate w/ Lid
Each for $5,670.61
Only null left
N16308S
View Documents Promotion Details
Thermo Scientific™
N16308S
Liquid Protection Tray
Each for $115.00
Only null left
FEVLUPLA06
View Documents Promotion Details
Thermo Scientific™
VLUPLA06
Upgrade kit for luminometry and AlphaScreen
Each for $4,845.24
Only null left
FEVLUPLT07
View Documents Promotion Details
Thermo Scientific™
VLUPLT07
Upgrade Kit for Luminometry and TRF
Each for $5,604.53
Only null left
FEVLUPLAT08
View Documents Promotion Details
Thermo Scientific™
VLUPLAT08
Upgrade kit for luminometry, AlphaScreen and TRF
Each for $6,413.72
Only null left
FEVLUPD209
View Documents Promotion Details
Thermo Scientific™
VLUPD209
Upgrade kit for two dispensers
Each for $6,486.61
Only null left
Description
Description
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.
Varioskan LUX is a modular instrument, you can upgrade it with missing features later.
  • Installation of the upgrade kits must be carried out by trained and authorized service personnel.
  • For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
  • TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
  • Please contact your local representative to discuss about the instrument upgrade option
Plate adapters are used for adapting plates that are of different heights.
  • Adapters adjust the plates to the optimum height for measuring and dispensing.
  • Choose the adapter based on the plate format in use and whether the lid is needed or not.
There are two types of plate trays: universal and robotic.
  • The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
  • A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
Specifications
Specifications
Varioskan