missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Accessories for Valor 3000 Xtreme Portable Food Portioning Scales

Catalog No. p-3585962
Change view
Click to view available options
Type:
Carrying Case
French Fry Holder
Ice Cream Cone Holder
Sandwich Pan
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01919400 Carrying Case
01919401 Sandwich Pan
01919402 French Fry Holder
01919403 Ice Cream Cone Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01919400 Supplier OHAUS™ Supplier No. 80251216
Only null left

For use with Valor 3000 Xtreme Portable Food Portioning Scales.

Sandwich Pan

  • Stainless steel
  • For weighing sandwiches and other long food items
  • Clips over the platform of the Valor 3000 Xtreme
  • Measures 11W x 5.9D x 0.4H in. (28 x 15 x 1cm)

French Fry Holder

  • Stainless steel
  • For weighing french fries, onion rings, tacos, and other food service items that are required to remain upright during weighing
  • Clips over the platform of the Valor 3000 Xtreme
  • Dimensions, front wall: 4.75 in. (12.1cm) wide by 1.13 in. (2.9cm) high; back wall: 4.75 in. (12.1cm) wide by 2.75 in. (7cm) high

Ice Cream Cone Holder

  • Stainless steel
  • Used for weighing ice cream, frozen custard, gelati, and other food service items where the cone has to be held to keep the item upright
  • Clips over the platform of the Valor 3000 Xtreme
  • Upper platform measures 5.9 in. (15cm) wide by 3.5 in. (8.9cm) deep
  • There are two holes in the upper platform, one 2.25 in. (5.7cm) and the other 1.5 in. (3.8cm)
  • Height of upper platform is 4 in. (10.2cm) above lower platform

Specifications

For Use With (Equipment) Valor 3000 Xtreme Digital Compact Scale
Type Carrying Case
Autoclavable Not Autoclavable