missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Accessories for Valor 3000 Xtreme Portable Food Portioning Scales

For use with Valor 3000 Xtreme Portable Food Portioning Scales.

Supplier:  OHAUS™ 80251149

 View more versions of this product

Catalog No. 01919403


Only null left

Description

Description

Sandwich Pan
  • Stainless steel
  • For weighing sandwiches and other long food items
  • Clips over the platform of the Valor 3000 Xtreme
  • Measures 11W × 5.9D × 0.4H in. (28 × 15 × 1cm)
French Fry Holder
  • Stainless steel
  • For weighing french fries, onion rings, tacos, and other food service items that are required to remain upright during weighing
  • Clips over the platform of the Valor 3000 Xtreme
  • Dimensions, front wall: 4.75 in. (12.1cm) wide by 1.13 in. (2.9cm) high; back wall: 4.75 in. (12.1cm) wide by 2.75 in. (7cm) high
Ice Cream Cone Holder
  • Stainless steel
  • Used for weighing ice cream, frozen custard, gelati, and other food service items where the cone has to be held to keep the item upright
  • Clips over the platform of the Valor 3000 Xtreme
  • Upper platform measures 5.9 in. (15cm) wide by 3.5 in. (8.9cm) deep
  • There are two holes in the upper platform, one 2.25 in. (5.7cm) and the other 1.5 in. (3.8cm)
  • Height of upper platform is 4 in. (10.2cm) above lower platform
Specifications

Specifications

Valor 3000 Xtreme Digital Compact Scale
5.9 in.
4 in.
Ice Cream Cone Holder
Stainless Steel
15 cm
10.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions