missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACAT2 Recombinant Rabbit Monoclonal Antibody (23GB4745)
GREENER_CHOICE

Catalog No. MA553804
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553804 100 μL
MA553805 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553804 Supplier Invitrogen™ Supplier No. MA553804
Only null left

Rabbit Recombinant Monoclonal Antibody

ACAT2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene represents a member of the 'frizzled' gene family, which encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The protein encoded by this family member contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, and seven transmembrane domains, but unlike other family members, this protein does not contain a C-terminal PDZ domain-binding motif. This protein functions as a negative regulator of the canonical Wnt/beta-catenin signaling cascade, thereby inhibiting the processes that trigger oncogenic transformation, cell proliferation, and inhibition of apoptosis. Alternative splicing results in multiple transcript variants.

Specifications

Antigen ACAT2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4745
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Acat2
Gene Accession No. Q5XI22, Q8CAY6, Q9BWD1
Gene Alias Ab2-076; Acat2; Acat3; acetoacetyl Coenzyme A thiolase; acetyl-CoA acetyltransferase 2; acetyl-CoA acetyltransferase, cytosolic; Acetyl-CoA transferase-like protein; acetyl-Coenzyme A acetyltransferase 2; acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase); acetyl-Coenzyme A acetyltransferase 3; ACTL; AE1; AW742799; BND3; CD233; cytosolic acetoacetyl-CoA thiolase; DI; EMPB3; EPB3; FR; RTA1A; SW; t-complex protein 1, related sequence 1; Tcp1-rs1; Tcp-1x; WD; WD1; WR
Gene Symbols Acat2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACAT2 (200-397AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110460, 308100, 39
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less