missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACAP2 Recombinant Rabbit Monoclonal Antibody (23GB1565)
GREENER_CHOICE

Catalog No. MA553176
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553176 100 μL
MA553177 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553176 Supplier Invitrogen™ Supplier No. MA553176
Only null left

Rabbit Recombinant Monoclonal Antibody

ACAP2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

ACAP2 is a transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain, and a PH domain. ACAP2, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6) has robust constitutive ARF6 GAP activity, where the GAP activity is stimulated by phosphatidylinositol 4,5-bisphosphate (PIP2) and phosphatidic acid but has little activity toward ARF1 and is known to control the return of ARF to the inactive GDB-bound shots. ACAP2, along with ACAP1, is targeted by phosphatidyl inositol (3, 4, 3) triphosphate inhibiting the formation of PDGF-induced dorsal membrane ruffles in NIH3T3 fibroblasts when overexpressed. ACAP2 is phosphorylated upon DNA damage, probably by ATM or ATR. Expression is wide-spread with the highest level in lung.

Specifications

Antigen ACAP2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1565
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACAP2
Gene Accession No. Q15057, Q5FVC7, Q6ZQK5
Gene Alias 4832442G16; 9530039J15Rik; ACAP2; Arf GAP with coiled coil, ANK repeat and PH domains 2; arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2; ArfGAP with coiled-coil, ankyrin repeat and PH domains 2; centaurin beta2; centaurin, beta 2; centaurin-beta-2; Centb2; CNT-B2; KIAA0041; mKIAA0041
Gene Symbols ACAP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACAP2 (650-720AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23527, 619382, 78618
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less