missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ AC Ammeter

Ideal option for classroom electricity demonstrations.

Supplier:  United Scientific™ MACA02

 View more versions of this product

Catalog No. S15793

Only null left

Description

Description

  • AC ammeter mounted in a black plastic case.
  • Scale length of 75 mm, with ± 2.5% accuracy
  • Features 5-way binding post and an external zero-setting screw
  • Not protected against prolonged or significant overload
  • Stackable for convenient storage
Specifications

Chemical Identifiers

Analog
Ammeter, Analog
Classroom Demonstrations
1 Ea.

Specifications

Classroom Demonstrations
1 Ea.
0 to 1 A/0 to 5 A
Ammeter, Analog
Analog
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Warning: Cancer and reproductive harm