missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Cylindrical Cells

Fit standard spectrophotometer cell holders

Supplier:  Fisherbrand™ 14958146

 View more versions of this product

Catalog No. 14958146



Only null left

Includes: A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).

Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 22mm wide
  • Available size is macro
  • 10, 20, 50 or 100mm path length rectangular cells
  • Available with a variety of window material for wide range of assays
  • Available in two materials e.g. Quartz and Special Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

Absorption Cylindrical Cell
16,000 μL
Special Optical Glass
320 to 2500 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions