missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Cylindrical Cells

Catalog No. p-4757371
Change view
Click to view available options
Capacity (Metric):
16,000 μL
31,000 μL
6300 μL
Material:
Optical Glass
Quartz
Special Optical Glass
Wavelength Range:
190 to 2600 nm, 2850 to 3600 nm
320 to 2500 nm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Wavelength Range
14958147 31,000 μL Optical Glass 320 to 2500 nm
14958146 16,000 μL Special Optical Glass 320 to 2500 nm
14958143 31,000 μL Quartz 190 to 2600 nm, 2850 to 3600 nm
14958142 16,000 μL Quartz 190 to 2600 nm, 2850 to 3600 nm
14958145 6300 μL Optical Glass 320 to 2500 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14958147 Supplier Fisherbrand™ Supplier No. 14958147
Only null left

Includes

A filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).

Fit standard spectrophotometer cell holders

  • All cells are 22mm wide
  • Available size is macro
  • 10, 20, 50 or 100mm path length rectangular cells
  • Available with a variety of window material for wide range of assays
  • Available in two materials e.g. Quartz and Special Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Type Cell
Capacity (Metric) 31,000 μL
Material Optical Glass
Width (Metric) Interior 18.5mm
Path Length 100 mm
Wavelength Range 320 to 2500 nm
Height (Metric) 35 mm
Width (Metric) 22 mm
Shape Cylinder