Learn More
Agilent BioTek Absorbance Test Plate
GREENER_CHOICE
For use with any microplate reader with 340 nm absorbance measurement capability to evaluate optical systems at 340 nm
Supplier:  Agilent BioTek 7260551
Catalog No. BT7260551
Only null left
Description
Description
  • Tests linearity, accuracy, repeatability and alignment
  • Includes six filters for testing 340 nm at different optical densities
  • With data sheet and instructions
Specifications
Specifications
Absorbance Test Plate
Agilent BioTek Microplate Readers
340 nm
for testing 340nm at different optical densities
Product Suggestions
Product Suggestions