missing translation for 'onlineSavingsMsg'
Learn More

(+)-Abscisic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013536
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013536 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013536 Supplier Merck Emd Millipore Supplier No. 9076925MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 21293-29-8
Linear Formula C15H20O4
MDL Number MFCD00066545
Synonym -5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2 Z,4 E)-pentadienoic acid; ABA; Dormin
Type Analytical Standard
Quantity 25 mg
Formula Weight 264.32
Percent Purity ≥98% (HPLC)
Grade Analytical Standard