missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abraxas Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Specifications
| Antigen | Abraxas |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt.
CCDC98, also known as Abraxas 1, was identified through protein binding studies using the breast and ovarian predisposition protein BRCA1 as the binding target. CCDC98 recruits RAP80, a ubiquitin-binding protein, to BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both CCDC98 and RAP80 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either CCDC98 or RAP80 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that CCDC98 and RAP80 control only part of the DNA damage response role of BRCA1. At least two isoforms of CCDC98 are known to exist.Specifications
| Abraxas | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| UNQ496/PRO1013, ABRA1, CCDC98, FLJ11520, FLJ12642, FLJ13614, BRCA1-A complex subunit Abraxas, abraxas protein, coiled-coil domain containing 98, coiled-coil domain-containing protein 98 | |
| ABRAXAS1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q6UWZ7 | |
| 84142 | |
| Between 350 and 400 | |
| Primary | |
| 4° C | |
| ABRAXAS1 |