missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ WST-1 Cell Proliferation Assay Kit

Catalog No. 89024504
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89024504 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89024504 Supplier Abnova™ Supplier No. KA1384

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • WST-1 reagent (powder), 1 vial/1 ea.
  • Electron mediator solution, 1 vial/1.2mL

WST-1 Cell Proliferation Assay Kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model.

  • The assay is based on the enzymatic cleavage of the tetrazolium salt WST-1 to formazan by cellular mitochondrial dehydrogenases present in viable cells
  • This kit will also allow investigators to screen drug candidates involved in regulation of cell cycle
  • This product is for laboratory research use only: not for administration to humans
  • Store at -20°C

Specifications

Description WST-1 Cell Proliferation Assay Kit provides an easy to use tool for studying induction and inhibition of cell proliferation in any in vitro model.
Product Type WST-1 cell proliferation assay kit
For Use With (Application) Functional Study
Kit Contents Electron mediator solution, WST-1 reagent
Quantity Each
Storage Requirements Store the kit at -20°C