missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Serotonin ELISA Kit

Catalog No. 89100615
Change view
Click to view available options
Sample Type:
Biological sample
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89100615 Biological sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89100615 Supplier Abnova™ Supplier No. KA2518
Only null left

Enzyme immunoassay for the quantitative determination of Serotonin in serum, urine and platelets

  • Sample Type: Platelet, Serum, Urine
  • Sample Volume: 25μL

Applications:

Quantification

Specifications

Description Serotonin ELISA Kit is an ultra-sensitive Enzyme Immunoassay for the quantitative determination of Serotonin.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Biological sample
Sample Volume 100 μL
Content And Storage 4°C
For Use With (Application) Quantification
Product Type Serotonin ELISA kit