missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ LCN2 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human NGAL/Lipocalin-2 in serum, plasma, urine, tissue culture medium, and other biological media.
$994.44 - $994.44
Specifications
| Conjugate | HRP |
|---|---|
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Catalog Number | Mfr. No. | Sample Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sample Type | Price | Quantity | |||||
|
89-100-014
|
Abnova™
KA0043 |
Plasma (Citrate, EDTA, Heparin), Serum, Urine |
Each for $994.44
|
|
|||||
Specifications
| HRP | |
| Colorimetric | |
| For research use only (RUO) | |
| 96 |
| Human | |
| 1 Kit | |
| Quantification |