missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL15 (Human) ELISA Kit
Use to measure quantitatively human IL15. Sandwich enzyme immunoassay.
Supplier: Abnova™ KA0376
Includes:
- 96-well plate precoated with anti- human IL-15 antibody, 96 (8 x 12) wells
- Lyophilized recombinant human IL-15 standard, 10ng/tube x 2
- Biotinylated anti-human IL-15 antibody, dilution 1:100, 130μL
- Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
- Sample diluent buffer, 30mL
- Antibody diluent buffer, 12mL
- ABC diluent buffer, 12mL
- TMB color developing agent, 10mL
- TMB stop solution, 10mL
Description
IL15 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technologySpecifications
| 15.6 to 1000pg/mL | |
| Human | |
| 3pg/mL | |
| 1 Kit | |
| Cell Culture Supernatant, Plasma (EDTA, heparin), Serum | |
| <3 pg/mL | |
| Store the kit at -20°C | |
| ELISA Kit | |
| 96 |
| Biotin | |
| IL15 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL15. | |
| Colorimetric | |
| For research use only (RUO) | |
| 100 μL | |
| 96-well | |
| Quantification | |
| No detectable cross-reactivity with any other cytokine |