missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Histamine ELISA Kit

Catalog No. p-4394539
Change view
Click to view available options
Conjugate:
AP
Peroxidase
Sample Volume:
25 μL (acylated samples)
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Sample Volume
89024540 AP 50 μL
89100719 Peroxidase 25 μL (acylated samples)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89024540 Supplier Abnova™ Supplier No. KA1420
Only null left

Includes

  • 96-well Microtiter plate (No. S): Twelve strips of 8 detachable wells, coated with Anti-Histamine antibody, 96 (8 x 12) wells
  • Calibrator: 0, 2.5, 5.0 and 10.0μg/mL of histamine. 0.9mL x 4
  • Histamine-Alkaline Phosphatase conjugate (HTM-ALP) (No. 3) 10.5mL
  • p-Nitrophenyl Phosphate (pNPP) substrate (No. 5). Ready to use. 10.5mL
  • Wash Buffer (10X PBS-Tween) (No. 6). Dilute 10 fold with distilled or deionized water to 150mL prior to use, 15mL
  • Stop Solution (No. 7): 3N NaOH. 5.5mL

Competitive binding Enzyme Immunoassay for the quantitative measurement of Histamine

Specifications

Conjugate AP
Cross Reactivity Fish
Description Histamine ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of histamine in fish.
Detection Method Colorimetric
Includes 12 Strip of 8 detachable well, 96-well microtiter plate, 1.2mL negative calibrator, 3 vials x 0.9mL positive calibrator, 10.5mL Histamine-Alkaline Phosphatase Conjugate (HTM-ALP), 10.5mL p-Nitrophenyl Phosphate (pNPP) Substrate, 15mL Wash Buffer, 5.5mL stop solution
Quantity 1 Kit
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Histamine ELISA Kit