missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Histamine ELISA Kit
Description
Includes
- 96-well Microtiter plate (No. S): Twelve strips of 8 detachable wells, coated with Anti-Histamine antibody, 96 (8 x 12) wells
- Calibrator: 0, 2.5, 5.0 and 10.0μg/mL of histamine. 0.9mL x 4
- Histamine-Alkaline Phosphatase conjugate (HTM-ALP) (No. 3) 10.5mL
- p-Nitrophenyl Phosphate (pNPP) substrate (No. 5). Ready to use. 10.5mL
- Wash Buffer (10X PBS-Tween) (No. 6). Dilute 10 fold with distilled or deionized water to 150mL prior to use, 15mL
- Stop Solution (No. 7): 3N NaOH. 5.5mL
Specifications
Specifications
| Conjugate | AP |
| Cross Reactivity | Fish |
| Description | Histamine ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of histamine in fish. |
| Detection Method | Colorimetric |
| Includes | 12 Strip of 8 detachable well, 96-well microtiter plate, 1.2mL negative calibrator, 3 vials x 0.9mL positive calibrator, 10.5mL Histamine-Alkaline Phosphatase Conjugate (HTM-ALP), 10.5mL p-Nitrophenyl Phosphate (pNPP) Substrate, 15mL Wash Buffer, 5.5mL stop solution |
| Quantity | 1 Kit |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | Histamine ELISA Kit |