missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Hepatitis B surface antigen Ab ELISA Kit
Specifications
Specifications
| Calibration Range | 0 to 1000mIU/mL |
| Conjugate | HRP |
| Description | Hepatitis B surface antigen antibody ELISA Kit is a sandwich enzyme immunoassay for the measurement of antibodies to hepatitis B surface antigen. |
| Detection Limit | 10mIU/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 6mL HBsAg -Peroxidase solution, 1mL Anti-HBs positive control, 1.5mL HB negative control, 10mL TMB Substrate solution A, 10mL TMB Substrate solution B, 50mL Conc. washing solution D (20X), 10mL 2N Sulfuric acid |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| Show More |