missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent Dye 488-I Phalloidin

Catalog No. 89139567
Change view
Click to view available options
Quantity:
300 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89139567 300 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89139567 Supplier Abnova™ Supplier No. U0281

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorescence Dye 488-I with Phalloidin conjugate

Fluorescence Dye 488-I conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescence Dye 488-I is not affected by pH between 4-10. Compared to Alexa Fluor 488, Fluorescence Dye 488-I of single isomer has much higher purity, bringing better performance consistency from batch to batch. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.

  • Spectral Equivalent: Alexa Fluor 488, Cy2, DyLight 488, FITC
  • Molar Extinction Co-efficient: ≥70,000 M-1cm-1
  • Excitation (Max): 491nm
  • Emission (Max): 514nm
  • Solubility: DMSO
  • Storage Buffer: DMSO

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye 488-I with Phalloidin conjugate. Fluorescence Dye 488-I conjugates are significantly brighter than fluorescein conjugates and much more photostable. Additionally, the fluorescence of Fluorescence Dye 488-I is not affected by pH between 4-10. Compared to Alexa Fluor 488, “Fluorescence Dye 488-I of single isomer has much hihger purity, bringing better performance consistancy from batch to batch.
Format Liquid
For Use With (Application) Conjugation
Quantity 300 Reactions
Product Type Fluorescent Dye 488-I Phalloidin
Sufficient For 300 Reactions
Equivalence Alexa Fluor 488, Cy2, DyLight 488, FITC
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 488-I Phalloidin
Label Type Other Labels or Dyes
Emission 514
Excitation 491
Show More Show Less

For research use only (RUO).