Learn More
Abnova™ Epinephrine/Norepinephrine ELISA Kit
Enzyme Immunoassay for the quantitative determination of Epinephrine and Norepinephrine in Plasma and Urine
Supplier:  Abnova™ KA3768
 View more versions of this product
Catalog No. 89-101-106
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Epinephrine: 0.5ng/mL to 80ng/mL, Norepinephrine: 0.2ng/mL to 32ng/mL
Colorimetric
Biological Fluid
Store the kit at 4°C
Epinephrine/Norepinephrine ELISA kit
Epinephrine/Norepinephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of Epinephrine and Norepinephrine in biological fluids.
1 Kit
1 to 750 μL
Quantification
Product Suggestions
Product Suggestions