missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Epinephrine/Norepinephrine ELISA Kit
Enzyme Immunoassay for the quantitative determination of Epinephrine and Norepinephrine in Plasma and Urine
Supplier: Abnova™ KA1877
Specifications
| Epinephrine: 1.3 to 81.4ng/mL, Norepinephrine: 4.9 to 339ng/mL | |
| Colorimetric | |
| 1 Kit | |
| 10 μL (Urine), 300 μL (Plasma) | |
| Quantification |
| Epinephrine/Norepinephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of Epinephrine and Norepinephrine in plasma and urine. This product contains two assay plates, for Epinephrine and Normetanephrine separately. | |
| Adhesive foil 4 x 2, 2 x 20mL Wash Buffer concentrate, 2 12mL Enzyme Conjugate, 2 12mL Substrate, 2 12mL stop solution, 96-well Adrenaline, 96-well Noradrenaline, 6mL Adrenaline antiserum from Rabbit, 6mL Noradrenaline antiserum from Rabbit, 4mL Adjustment Buffer, 4mL standard A, 4mL standard B, 4mL, standard C, 4mL standard D, 4mL standard E, 4mL standard F, 20mL Acylation Buffer, 3mL Acylation reagent, 6mL Assay Buffer, 4mL coenzyme, four 1mL Enzyme lyophilized, 6mL Extraction Buffer, 2 48-well extraction plate coated with boronate affinity gel, 20mL Hydrochloric acid, 4mL control 1, 4mL control 2 | |
| Plasma, Urine | |
| Store the kit at 4°C | |
| Epinephrine/Norepinephrine ELISA kit |