missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Dopamine ELISA Kit

Catalog No. 89028620
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89028620 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89028620 Supplier Abnova™ Supplier No. KA1887

Includes

  • Adhesive foil ready for use, 1 x 4
  • Wash buffer concentrate, Concentrate. Dilute content with dist. Water to a final volume of 1000mL, 1 x 20mL
  • Enzyme conjugate ready for use, anti-rabbit IgG conjugated with peroxidase, 1 x 12mL
  • Substrate ready for use, containing a solution of tetramethylbenzidine (TMB), 1 x 12mL
  • Stop solution ready for use, containing 0.25M H2SO4, 1 x 12mL
  • Dopamine microtiter Strips 12 strips, 8 wells each, break apart, pre-coated, green coloured, 1 x 96 wells
  • Dopamine antiserum from rabbit, ready for use, green coloured, green screw cap, 1 x 6mL
  • Adjustment buffer ready for use, 1 x 4mL
  • Standard A ready for use, 1 x 4mL
  • Standard B ready for use, 1 x 4mL
  • Standard C ready for use, 1 x 4mL

Enzyme Immunoassay for the quantitative determination of Dopamine in Plasma and Urine

Specifications

Calibration Range 114 to 1917ng/mL Urine, 1000 to 8000pg/mL Plasma
Description Dopamine ELISA Kit is an enzyme immunoassay for the quantitative determination of Dopamine in plasma and urine.
Detection Method Colorimetric
Includes 20mL Wash Buffer concentrate, 12mL Enzymeconjugate, 12mL Substrate, 12mL stop solution, 96-Dopamine microtiter strip, 6mL Dopamine antiserum, 4mL Adjustment Buffer, 4mL standard A, 4mL standard B, 4mL standard C, 4mL standard D, 4mL standard E, 4mL standard F, 20mL Acylation Buffer, 3mL Acylation reagent, 6mL Assay Buffer, 4mL coenzyme, 2 1mL Enzyme, 6mL Extraction Buffer, 2 48-well extraction plate, 20mL Hydrochloric acid, 4mL control 1, 4mL control 2, 4mL standard A/B
Quantity 1 Kit
Sample Type Plasma, Urine
Sample Volume Urine: 10 μL, Plasma: 300 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Dopamine ELISA kit