missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cortisol ELISA Kit

Catalog No. 89028618
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89028618 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89028618 Supplier Abnova™ Supplier No. KA1885

Enzyme Immunoassay for the quantitative determination of Cortisol in Human Saliva

Specifications

Description Cortisol ELISA Kit is an enzyme immunoassay for the quantitative determination of Cortisol in human saliva.
Detection Limit 0 to 30ng/mL
Detection Method Colorimetric
Includes 96-well microtiter coated (12 x 8 strip) with Anti-cortisol Antibody (Polyclonal), 26mL Enzyme Conjugate Cortisol Conjugated to HRP, 1mL Cortisol standard A, 0ng/ml, 1mL Cortisol standard B, 0.1ng/m, 1mL Cortisol standard C, 0.5ng/m, 1mL Cortisol standard D,, 1.5ng/ml, 1mL Cortisol standard E, 4ng/ml, 1mL Cortisol standard F, 10ng/ml, 1mL Cortisol standard G, 30ng/ml, 1mL/vial, 2 vials Control 1 and 2, 30mL wash solution (40X concentrated), 25mL Substrate solution, 14mL stopping solution
Quantity 1 Kit
Sample Type Saliva
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cortisol ELISA kit