missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cortisol ELISA Kit

For the quantitative measurement of free Cortisol in Urine

$632.31 - $632.31

Specifications

Conjugate HRP
Detection Method Colorimetric
Regulatory Status For research use only (RUO)
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
View More Specs

Products 1
Catalog Number Mfr. No. Sample Type Price Quantity  
Catalog Number Mfr. No. Sample Type Price Quantity  
89-100-919
View Documents
Abnova™
KA3382
Dried Fecal Extract, Plasma, Saliva, Serum, Tissue Culture Medium Sample, Urine
Each for $632.31
Only null left
 
Description

Description

Specifications

Specifications

HRP
For research use only (RUO)
Quantification
Colorimetric
Store the kit at 4°C
96
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'