Learn More
Abnova™ Corticosterone (Human) ELISA Kit
Supplier: Abnova™ KA0468
Specifications
| 0.391 to 100ng/mL | |
| Colorimetric | |
| Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine |
| Biotin | |
| 1 Kit | |
| 25 μL |