missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Bdnf (Mouse) ELISA Kit

Catalog No. 89022808
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89022808 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89022808 Supplier Abnova™ Supplier No. KA0331

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Lyophilized recombinant mouse BDNF standard 10ng/tube+2
  • One 96-well plate precoated with anti- mouse BDNF antibody 1
  • Sample diluent buffer 30mL
  • Biotinylated anti- mouse BDNF antibody, dilution 1:100 130μL
  • Antibody diluent buffer 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100 130μL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Bdnf

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Bdnf (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Bdnf.
Detection Limit 2pg/mL
Detection Method Colorimetric
Includes 96-well plate precoated with anti Mouse BDNF Antibody 96 (8x12) well, lyophilized recombinant Mouse BDNF standard 10ng/tube x 2, 130μL Biotinylated Anti-Mouse BDNF Antibody (dilution 1:100), 130μL Avidin Biotin Peroxida
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type BDNF (Mouse) ELISA kit
Show More Show Less