missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ADIPOQ (Human) ELISA Kit
For detection of ADIPOQ in Human Urine and Murine, Adiponectin in Plasma, Serum and cell culture supernatants
$1256.94 - $1256.94
Specifications
| Detection Method | Colorimetric |
|---|---|
| Quantity | 96 Assays |
| Regulatory Status | For research use only (RUO) |
| For Use With (Application) | Quantification |
| Product Type | ADIPOQ (Human) ELISA Kit |
| Catalog Number | Mfr. No. | Sample Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Sample Type | Price | Quantity | |||||
|
89-100-012
|
Abnova™
KA0017 |
CSF, Plasma (Citrate, EDTA, Heparin), Serum, Urine |
Each for $1,256.94
|
|
|||||
|
89-100-013
|
Abnova™
KA0016 |
Plasma (Citrate, EDTA, Heparin), Serum |
Each for $1,256.94
|
|
|||||
Specifications
| Colorimetric | |
| For research use only (RUO) | |
| ADIPOQ (Human) ELISA Kit |
| 96 Assays | |
| Quantification | |
| 96 |