Learn More
Applied Biosystems™ ABI LV40™ dAbz Pack
Catalog No. 4316341
Encompass
Change view
Click to view available options
Quantity:
60 Columns
Catalog No. Quantity
4316341 60 Columns
1 options
Catalog No. 4316341 Supplier Applied Biosystems™ Supplier No. 4316341
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Contains 60 dA(bz) ABI LV40™ columns.The dA(bz) ABI LV40™ columns are fully assembled, low-cost columns that produce high-quality oligonucleotides. ABI columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.
Save Time and MoneyCompared to conventional columns, the ABI LV40™ columns reduce cycle time by about 10% and the cost of synthesis by about 25%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
  • Proprietary design and manufacturing methods ensure consistency and high quality
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
  • Hydrophobic polystryrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis
  • The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length
Specifications
Packaging Type Shipped at Room Temperature
Quantity 60 Columns
For Research Use Only. Not for use in diagnostic procedures.