missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCD1 Recombinant Rabbit Monoclonal Antibody (23GB3125)
GREENER_CHOICE

Catalog No. MA553354
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553354 100 μL
MA553355 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553354 Supplier Invitrogen™ Supplier No. MA553354
Only null left

Rabbit Recombinant Monoclonal Antibody

ABCD1 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen ABCD1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3125
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ABCD1
Gene Accession No. P33897, P48410
Gene Alias ABC42; ABCD1; Adrenoleukodystrophy protein; ALD; Aldgh; ALDP; AMN; ATP binding cassette subfamily D member 1; ATP-binding cassette sub-family D member 1; ATP-binding cassette, subfamily D (ALD), member 1; ATP-binding cassette, sub-family D (ALD), member 1; ATP-binding cassette, sub-family D, member 1; RGD1562128; X-linked adrenoleukodystrophy (ALD) gene homolog
Gene Symbols ABCD1
Host Species Rabbit
Immunogen A synthetic peptide of human ABCD1/ALD (650-745AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11666, 215, 363516
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less