missing translation for 'onlineSavingsMsg'
Learn More

Abcam VHH Single Domain Anti-Rabbit IgG Fc, 250 uL

Catalog No. 502657497
Encompass
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502657497 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502657497 Supplier Abcam Inc Supplier No. AB218695250UL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Llama Monoclonal antibody, suitable for IHC-P, WB, ELISA, with 24 citations.

  • Host Species: Llama
  • Clonality: Monoclonal
  • Applications: IHC-P, WB, ELISA
  • Species Reactivity: Rabbit
  • Storage Buffer: Constituents: PBS, Sodium chloride
  • Form: Liquid
  • Purification: Affinity purification His Tag

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GD001
Conjugate Unconjugated
Formulation Constituents: PBS, Sodium chloride
Gene Rabbit IgG
Host Species Llama
Purification Method Affinity purification His Tag
Quantity 250 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Rabbit.
Target Species Rabbit
Product Type Monoclonal Antibody
Form Liquid
Host abrv Ll
Shipping Condition Blue Ice
Reactivity abrv Rb
Target Molecule Rabbit IgG
Validated Application abrv IHC-P, WB, ELISA
Show More Show Less