missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [SB76b] Anti-Mouse IgG3 gamma chain (Alexa Fluor™ 647), 250 ug

Catalog No. 502612919
Encompass
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502612919 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502612919 Supplier Abcam Inc Supplier No. AB172329250UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal antibody, suitable for ELISA, IHC-P, ICC/IF, Flow Cyt.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG3
  • Applications: ELISA, IHC-P, ICC/IF, Flow Cyt
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
  • Form: Liquid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ig gamma-3 chain C region
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SB76b
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Gene Ig gamma-3 chain C region
Host Species Rat
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG3
Host abrv Rt
Shipping Condition Blue Ice
Validated Application abrv ELISA, IHC-P, ICC/IF, Flow Cyt
Show More Show Less