missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rat monoclonal [LO-MG2b-1] Anti-Mouse IgG2b heavy chain, 250 ug

Catalog No. 502614480
Encompass
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502614480 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502614480 Supplier Abcam Inc Supplier No. AB11566250UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal antibody, suitable for ELISA, with 1 citations.

  • Host Species: Rat
  • Clonality: Monoclonal
  • Isotype: IgG1
  • Applications: ELISA
  • Species Reactivity: Mouse
  • Storage Buffer: Preservative: 0.09% Sodium azide, Constituents: PBS
  • Form: Liquid
  • Purification: Affinity purification Protein G

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications ELISA
Classification Monoclonal
Clone LO-MG2b-1
Formulation Preservative: 0.09% Sodium azide, Constituents: PBS
Gene Mouse IgG2b
Host Species Rat
Purification Method Affinity purification Protein G
Quantity 250 μg
Regulatory Status RUO
Whole Molecule Yes
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Mouse IgG2b
Validated Application abrv ELISA
Show More Show Less