missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AbC™ Total Antibody Compensation Bead Kit
Provides a consistent, accurate, and simple-to-use technique for the setting of flow cytometry compensation when using fluorochrome-conjugated mouse, rat, hamster, or rabbit antibodies with any laser in flow cytometry.
Supplier: Invitrogen™ A10513
Description
- Broadest multispecies reactivity—for use with all isotypes of mouse, rat, hamster, and rabbit antibody conjugates
- Brightest signal—most sensitive antibody-capture beads available
- Maximum laser compatibility—for use with all lasers
- Each kit provides polystyrene microspheres that have fluorescent antibody conjugate capture capacity (positive compensation beads) or are inert (negative compensation beads).
- Ideal for compensation with all isotypes of mouse (IgG1, IgG2a, IgG2b, IgG3), rat (IgG1, IgG2a, IgG2b), hamster (Armenian IgG and Syrian IgG), and rabbit (monoclonal and polyclonal) immunoglobulins.Demonstrates the highest and most consistent reactivity to the different subclasses of immunoglobulin in the industry.
Specifications
| 6 μm | |
| Tube | |
| 25 Tests | |
| Fluorescence | |
| Room Temperature | |
| Compensation Beads |
| Contains 1 vial AbC™ Total Compensation capture beads and 1 vial of negative beads. |
|
| Flow Cytometer | |
| 25 tests | |
| any lasers (UV to 633/635) | |
| AbC™ |
For Research Use Only. Not for use in diagnostic procedures.