missing translation for 'onlineSavingsMsg'
Learn More

Abacavir sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013404
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013404 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013404 Supplier Merck Emd Millipore Supplier No. PHR1256500MG
Abacavir sulfate belongs to the class of human immunodeficiency virus (HIV) medicines called nucleoside reverse transcriptase inhibitors, with antiretroviral activity against HIV.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 188062-50-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C28H36N12O2 · H2 SO4
Synonym Abacavir sulfate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 670.74
Grade Certified Reference Material