missing translation for 'onlineSavingsMsg'
Learn More

Abacavir Impurity Standard, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022860
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022860 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022860 Supplier Merck Emd Millipore Supplier No. BP1113

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 188062-50-2
Molecular Formula C28H36N12O2 · H2SO4
Linear Formula C28H36N12O2 · H2SO4
Synonym Abacavir sulfate
Type Pharmacopeia and Metrological Institutes Standards
Quantity 10 mg
Formula Weight 670.74
Grade British Pharmacopoeia (BP) Reference Standard