missing translation for 'onlineSavingsMsg'
Learn More

AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) 95.0+%, TCI America™

Catalog No. A5576100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5576100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5576100MG Supplier TCI America Supplier No. A5576100MG

Chemical Identifiers

CAS 254973-02-9
Molecular Formula C8H7N3O2S
Molecular Weight (g/mol) 209.223
MDL Number MFCD04038408
InChI Key JPARFDNKICYVCE-UHFFFAOYSA-N
Synonym 4-Acetamido-7-mercaptobenzofurazan, 4-Acetamido-7-mercapto-2,1,3-benzoxadiazole
PubChem CID 44630078
IUPAC Name N-(4-sulfanylidene-1H-2,1,3-benzoxadiazol-7-yl)acetamide
SMILES CC(=O)NC1=C2C(=NON2)C(=S)C=C1

Specifications

Quantity 100 mg
Formula Weight 209.22
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material AABD-SH (=4-Acetamido-7-mercapto-2,1,3-benzoxadiazole) [for HPLC Labeling]
TSCA No
Recommended Storage Refrigerator