accessibility menu, dialog, popup

UserName

CAS RN 254973-02-9

IUPAC Name: N-(7-sulfanylidene-3,7-dihydro-2,1,3-benzoxadiazol-4-yl)acetamide
Synonyms: N-(7-sulfanylidene-3H-2,1,3-benzoxadiazol-4-yl)acetamide
Molecular Weight (g/mol): 209.22
Molecular Formula: C8H7N3O2S
InChi Key: JPARFDNKICYVCE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C2NON=C2C(=S)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results