missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ A-431 Whole Cell Lysate

Catalog No. NB800PC12
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB800PC12 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB800PC12 Supplier R&D Systems™ Supplier No. NB800PC12

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

The A-431 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

Quantity 0.5 mL
Storage Buffer 1X SDS-PAGE Sample Buffer [0.05mM Tris HCl, 2% SDS, 10% Glycerol, 0.1% bromophenol blue, pH 6.8]
Purity Protein
Test Specificity A431 whole cell extract lysate
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.
Product Line Novus
For Use With (Application) Western Blot
Tissue Skin

For Research Use Only