missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glipizide, 98+%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Ignition Residue | 0.4% max. |
| Loss on Drying | 1% max. (100°C, 3 hrs) (vacuum) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Merck Index | 15, 4477 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in dmso and methanol |
| Formula Weight | 445.5 |
| Percent Purity | 98+% |
| Show More |
Safety and Handling
| EINECSNumber | 249-427-6 |
RUO – Research Use Only