missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9H-Pyrido[2,3-b]indole 98.0+%, TCI America™
Supplier: TCI America P23101G
Specifications
| 9H-Pyrido[2,3-b]indole | |
| 216°C | |
| C11H8N2 | |
| 1 g | |
| BPMFPOGUJAAYHL-UHFFFAOYSA-N | |
| 9H-pyrido[2,3-b]indole | |
| 67486 | |
| ≥98.0% (GC,T) |
| 244-76-8 | |
| 364°C | |
| MFCD00473709 | |
| alpha-Carboline | |
| C1=CC=C2C(=C1)C3=C(N2)N=CC=C3 | |
| 168.199 | |
| 168.20 | |
| Crystalline Powder |
Chemical Identifiers
| 244-76-8 | |
| 168.199 | |
| BPMFPOGUJAAYHL-UHFFFAOYSA-N | |
| 67486 | |
| C1=CC=C2C(=C1)C3=C(N2)N=CC=C3 |
| C11H8N2 | |
| MFCD00473709 | |
| alpha-Carboline | |
| 9H-pyrido[2,3-b]indole |
Safety and Handling
TSCA : No