accessibility menu, dialog, popup

UserName

HNN

CAS RN 244-76-8

IUPAC Name: 9H-pyrido[2,3-b]indole
Synonyms: 9H-pyrido[2,3-b]indole
Molecular Weight (g/mol): 168.2
Molecular Formula: C11H8N2
InChi Key: BPMFPOGUJAAYHL-UHFFFAOYSA-N
SMILES: N1C2=C(C=CC=C2)C2=C1N=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results