missing translation for 'onlineSavingsMsg'
Learn More

9H-Pyrido[2,3-b]indole 98.0+%, TCI America™

Catalog No. P2310200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2310200MG 200 mg
P23101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2310200MG Supplier TCI America Supplier No. P2310200MG
Only null left

Chemical Identifiers

CAS 244-76-8
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.199
MDL Number MFCD00473709
InChI Key BPMFPOGUJAAYHL-UHFFFAOYSA-N
Synonym alpha-Carboline
PubChem CID 67486
IUPAC Name 9H-pyrido[2,3-b]indole
SMILES C1=CC=C2C(=C1)C3=C(N2)N=CC=C3

Specifications

Melting Point 216°C
Boiling Point 364°C
Quantity 200 mg
Formula Weight 168.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9H-Pyrido[2,3-b]indole