Learn More
Greiner Bio-One 96-well Flat Bottom Chimney Style Non-binding Microplates
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well Flat Bottom Chimney Style Non-binding Microplates are free of detectable DNase, RNase, human DNA.
$957.43 - $963.10
Specifications
Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Non Binding
View More Specs
Products 2
Catalog Number Mfr. No. Well Color Price Quantity  
Catalog Number Mfr. No. Well Color Price Quantity  
07000634
View Documents
Greiner Bio-One
GB655900
Black N/A
Only null left
07000636
View Documents
Greiner Bio-One
655904
White N/A
Only null left
Description
Description
  • Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
  • Stable under normal assay conditions without degradation or leaching
  • Improves signal-to-noise-ratio
  • Manufactured under DIN ISO 9001 guidel
    Service Plan
    No
    Warranty
    No
Specifications
Specifications
No
Without Lid
Non Binding
96
Non-treated
Flat Bottom (F-well)
40/Cs.
Standard
8 x 12
Polystyrene
Non-sterile
392 μL
10
127.8 x 85.5 x 14.4 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'